2-Fluoro-5-nitrobenzotrifluoride ≥99%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene
002721-250G
002721-25G
002721-100G
101776-312EA
176.92
USD
101776-312
101776-308
101776-310
2-Fluoro-5-nitrobenzotrifluoride ≥99%
2-Fluoro-5-nitrobenzotrifluoride
Hazard: Toxic/Irritant
Formula:
C₇H₃F₄NO₂ Boiling Pt: 104…105 °C (20 mmHg) Density: 1.522 |
MDL Number:
MFCD00024576 CAS Number: 400-74-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 104-105°/2 |
Density | 1.522 |
ND20 | 1.465 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...