2-Fluoro-5-nitrobenzonitrile ≥97%

Supplier: Matrix Scientific

002715-25G 002715-100G
101776-300EA 73.85 USD
101776-300 101776-302
2-Fluoro-5-nitrobenzonitrile ≥97%
2-Fluoro-5-nitrobenzonitrile

Hazard: Toxic/Irritant

Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00042299
CAS Number: 17417-09-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 70-72°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR