2-Chloro-5-fluorotoluene ≥99%
Supplier: Matrix Scientific
002701-25G
002701-100G
101776-296EA
33.85
USD
101776-296
101776-298
2-Chloro-5-fluorotoluene ≥99%
2-Chloro-5-fluorotoluene
Hazard: Flammable
Formula:
C₇H₆ClF Boiling Pt: 156…157 °C Density: 1.2 Flash Pt: 51 °C |
MDL Number:
MFCD00040853 CAS Number: 33406-96-1 |
Specification Test Results
Boiling point | 156-157° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...