4-Chloro-3-(trifluoromethyl)benzonitrile
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-cyanobenzotrifluoride
002686-5G
002686-25G
101776-282EA
26.15
USD
101776-282
101776-284
4-Chloro-3-(trifluoromethyl)benzonitrile
4-Chloro-3-(trifluoromethyl)benzonitrile
Hazard: Toxic
Formula:
C₈H₃ClF₃N MW: 205.57 |
MDL Number:
MFCD00190117 CAS Number: 1735-54-2 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...