2-Fluoro-5-(trifluoromethyl)anisole

Supplier: Matrix Scientific

002653-5G 002653-1G
101776-266EA 120 USD
101776-266 101776-264
2-Fluoro-5-(trifluoromethyl)anisole
2-Fluoro-5-(trifluoromethyl)anisole

Hazard: Flammable

Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD01631564
CAS Number: 261951-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR