2-Chloro-5-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

002652-1G 002652-5G
101776-260EA 30.77 USD
101776-260 101776-262
2-Chloro-5-(trifluoromethyl)phenylacetonitrile
2-Chloro-5-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅ClF₃N
MW: 219.59 g/mol
MDL Number: MFCD01631472
CAS Number: 22902-88-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR