2-Chloro-5-(trifluoromethyl)benzoyl chloride

Supplier: Matrix Scientific

002649-5G 002649-1G
101776-258EA 138.46 USD
101776-258 101776-256
2-Chloro-5-(trifluoromethyl)benzoyl chloride
2-Chloro-5-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₃Cl₂F₃O
Boiling Pt: 59…61 °C (1 mmHg)
MDL Number: MFCD01631634
CAS Number: 657-05-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR