2-Chloro-5-(trifluoromethyl)anisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-methoxy-4-(trifluoromethyl)benzene

002647-1G 002647-5G
101776-252EA 32.31 USD
101776-252 101776-254
2-Chloro-5-(trifluoromethyl)anisole ≥97%
2-Chloro-5-(trifluoromethyl)anisole

Hazard: Flammable

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Boiling Pt: 100…101 °C (38 Torr)
MDL Number: MFCD01631470
CAS Number: 402-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR