2-Fluoro-5-(trifluoromethyl)phenylacetonitrile ≥98%

Supplier: Matrix Scientific

002638-1G 002638-5G
101776-238EA 29.23 USD
101776-238 101776-240
2-Fluoro-5-(trifluoromethyl)phenylacetonitrile ≥98%
2-Fluoro-5-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅F₄N
MDL Number: MFCD00061180
CAS Number: 220227-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR