3-Fluoro-5-(trifluoromethyl)phenol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-5-hydroxybenzotrifluoride

002635-1G 002635-5G
101776-234EA 29.23 USD
101776-234 101776-236
3-Fluoro-5-(trifluoromethyl)phenol ≥98%
3-Fluoro-5-(trifluoromethyl)phenol

Hazard: Corrosive

Formula: C₇H₄F₄O
MW: 180.1 g/mol
MDL Number: MFCD00061288
CAS Number: 172333-87-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR