2-Fluoro-5-(trifluoromethyl)phenol ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Fluor-2-hydroxy-4-(trifluormethyl)benzol, 2-Fluoro-5-trifluoromethylphenol

002634-5G 002634-25G 002634-100G
101776-228EA 27.69 USD
101776-228 101776-230 101776-232
2-Fluoro-5-(trifluoromethyl)phenol ≥98%
2-Fluoro-5-(trifluoromethyl)phenol

Hazard: Flammable/Irritant

Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 146 °C
Density: 1.436
Flash Pt: 57 °C
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Boiling point 146°
Density 1.436

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR