3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98%

Supplier: Matrix Scientific

002633-5G 002633-1G
101776-226EA 120 USD
101776-226 101776-224
3-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98%
3-Fluoro-5-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₅BrF₄
MW: 257.03
Boiling Pt: 43
MDL Number: MFCD00061175
CAS Number: 239087-09-3

Order Now

Specification Test Results

Boiling point 43°/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR