2-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-fluoro-5-(trifluoromethyl)benzene, 2-Fluoro-5-(trifluoromethyl)benzyl Bromide

002632-5G 002632-1G
101776-222EA 84.62 USD
101776-222 101776-220
2-Fluoro-5-(trifluoromethyl)benzyl bromide ≥98%
2-Fluoro-5-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₅BrF₄
MW: 257.03 g/mol
MDL Number: MFCD00061174
CAS Number: 220239-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR