3-Fluoro-5-(trifluoromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

002617-5G 002617-25G
101776-216EA 20 USD
101776-216 101776-218
3-Fluoro-5-(trifluoromethyl)benzonitrile ≥97%
3-Fluoro-5-(trifluoromethyl)benzonitrile

Hazard: Toxic

Formula: C₈H₃F₄N
Density: 1.35
Flash Pt: 65 °C
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR