2-Fluoro-5-(trifluoromethyl)benzonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-5-trifluoromethylbenzonitrile, 3-Cyano-4-fluorobenzotrifluoride
002615-5G
002615-25G
002615-100G
101776-210EA
26.15
USD
101776-210
101776-212
101776-214
2-Fluoro-5-(trifluoromethyl)benzonitrile ≥97%
2-Fluoro-5-(trifluoromethyl)benzonitrile
Hazard: Toxic
Formula:
C₈H₃F₄N MW: 189.11 g/mol Boiling Pt: 185-187°C |
MDL Number:
MFCD00061280 CAS Number: 4088-84-0 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 185-187° |
ND20 | 1.446 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...