3-Fluoro-4-methylbenzyl bromide

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-2-fluoro-1-methylbenzene

002578-1G 002578-5G
101776-198EA 43.08 USD
101776-198 101776-200
3-Fluoro-4-methylbenzyl bromide
3-Fluoro-4-methylbenzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₈BrF
MW: 203.05
MDL Number: MFCD01631534
CAS Number: 145075-44-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR