3-Fluoro-4-methylbenzylamine
Supplier: Matrix Scientific
002577-1G
002577-5G
101776-194EA
36.92
USD
101776-194
101776-196
3-Fluoro-4-methylbenzylamine
3-Fluoro-4-methylbenzylamine
Hazard: Corrosive
Formula:
C₈H₁₀FN MW: 139.17 g/mol |
MDL Number:
MFCD01631531 CAS Number: 261951-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...