2-Fluoro-p-toluidine ≥98%

Supplier: Matrix Scientific

002566-25G 002566-100G
101776-182EA 36.92 USD
101776-182 101776-184
2-Fluoro-p-toluidine ≥98%
2-Fluoro-p-toluidine

Hazard: Toxic/Irritant

Formula: C₇H₈FN
Boiling Pt: 70…71 °C (7 mmHg)
Density: 1.108
MDL Number: MFCD00040975
CAS Number: 452-80-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 70-71°/7mm
Density 1.108
ND20 1.533


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR