3-Fluoro-p-toluidine ≥99%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methylaniline, 4-Amino-2-fluorotoluene

002565-25G 002565-100G
101776-178EA 32.31 USD
101776-178 101776-180
3-Fluoro-p-toluidine ≥99%
3-Fluoro-p-toluidine

Hazard: Toxic/Irritant

Formula: C₇H₈FN
MW: 125.15
Boiling Pt: 97
Melting Pt: 37
Density: 1.093
MDL Number: MFCD00007762
CAS Number: 452-77-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Melting point 30-32°
Density 1.093
ND20 1.54


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR