3-Chloro-4-fluorophenylacetonitrile ≥97%
Supplier: Matrix Scientific
002519-5G
002519-1G
101776-174EA
55.38
USD
101776-174
101776-172
3-Chloro-4-fluorophenylacetonitrile ≥97%
3-Chloro-4-fluorophenylacetonitrile
Hazard: Toxic
Formula:
C₈H₅ClFN MW: 169.5834 g/mol Boiling Pt: 130…134 °C (7 Torr) |
MDL Number:
MFCD01631553 CAS Number: 658-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...