3-Chloro-4-fluorobenzoyl chloride
Supplier: Matrix Scientific
002508-5G
002508-25G
101776-164EA
38.46
USD
101776-164
101776-166
3-Chloro-4-fluorobenzoyl chloride
3-Chloro-4-fluorobenzoyl chloride
Hazard: Corrosive/Irritant
Formula:
C₇H₃Cl₂FO Boiling Pt: 222 °C |
MDL Number:
MFCD00236215 CAS Number: 65055-17-6 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 222° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...