1-Chloro-4-fluorobenzene ≥98%

Supplier: Matrix Scientific

002507-100G 002507-500G
101776-160EA 253.85 USD
101776-160 101776-162
1-Chloro-4-fluorobenzene ≥98%
1-Chloro-4-fluorobenzene

TCA listed: T
Hazard: Flammable

Formula: C₆H₄ClF
Boiling Pt: 129-130°C
Density: 1.226
MDL Number: MFCD00000603
CAS Number: 352-33-0

Order Now

Specification Test Results

Boiling point 129-130°
Density 1.226
ND20 1.495


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR