2-Chloro-4-fluorobenzenesulphonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-fluorobenzene-1-sulphonyl chloride

002506-1G 002506-5G
101776-156EA 23.08 USD
101776-156 101776-158
2-Chloro-4-fluorobenzenesulphonyl chloride ≥97%
2-Chloro-4-fluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₃Cl₂FO₂S
Boiling Pt: 108-110°C/0.1mm
Melting Pt: 28-31°C
MDL Number: MFCD00052957
CAS Number: 85958-57-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 108-110°/0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR