3-Chloro-4-fluorobenzylamine ≥97%

Supplier: Matrix Scientific

002505-5G 002505-25G
101776-152EA 33.85 USD
101776-152 101776-154
3-Chloro-4-fluorobenzylamine ≥97%
3-Chloro-4-fluorobenzylamine

Hazard: Corrosive/Air Sensitive/Refrigerate

Formula: C₇H₇ClFN
MW: 159.59 g/mol
Boiling Pt: 104-106°C/15mm
MDL Number: MFCD00042507
CAS Number: 72235-56-4

Order Now

Specification Test Results

Boiling point 104-106°/1
ND20 1.536

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR