3-Chloro-4-fluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-fluorobenzene-1-sulphonyl chloride

002504-5G 002504-25G 002504-100G
101776-146EA 15.38 USD
101776-146 101776-148 101776-150
3-Chloro-4-fluorobenzenesulfonyl chloride ≥97%
3-Chloro-4-fluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Boiling Pt: 104-105°C/1mm
MDL Number: MFCD00041444
CAS Number: 91170-93-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 104-105°/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR