α-Bromo-3-chloro-4-fluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-fluorobenzyl bromide, 4-(Bromomethyl)-2-chloro-1-fluorobenzene

002503-25G 002503-100G
101776-142EA 46.15 USD
101776-142 101776-144
α-Bromo-3-chloro-4-fluorotoluene ≥98%
α-Bromo-3-chloro-4-fluorotoluene

Hazard: Corrosive/Refrigerate

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 63…65 °C (0.6 mmHg)
MDL Number: MFCD01631551
CAS Number: 192702-01-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 63-65°/0.6
ND20 1.568

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR