α-Bromo-2-chloro-4-fluorotoluene

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

002502-25G 002502-100G
101776-138EA 49.23 USD
101776-138 101776-140
α-Bromo-2-chloro-4-fluorotoluene
α-Bromo-2-chloro-4-fluorotoluene

Hazard: Corrosive/Lachrymator

Formula: C₇H₅BrClF
MW: 223.47
Melting Pt: 33…35 °C
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Melting point 33-35°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR