3-Chloro-4-fluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-fluorobenzyl Alcohol, (3-Chloro-4-fluorophenyl)methanol

002501-25G 002501-5G
101776-136EA 89.23 USD
101776-136 101776-134
3-Chloro-4-fluorobenzyl alcohol ≥98%
3-Chloro-4-fluorobenzyl alcohol

Hazard: Irritant

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD01631550
CAS Number: 161446-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR