3-Chloro-4-fluoroaniline ≥98%

Supplier: Matrix Scientific

002491-100G 002491-500G
101776-126EA 32.31 USD
101776-126 101776-128
3-Chloro-4-fluoroaniline ≥98%
3-Chloro-4-fluoroaniline

TCA listed: T
Hazard: Toxic

Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 227…228 °C
Melting Pt: 44…47 °C
MDL Number: MFCD00007767
CAS Number: 367-21-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 227-228°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR