5-Chloro-4-fluoro-2-nitroaniline ≥97%

Supplier: Matrix Scientific

002486-1G 002486-5G
101776-118EA 21.54 USD
101776-118 101776-120
5-Chloro-4-fluoro-2-nitroaniline ≥97%
5-Chloro-4-fluoro-2-nitroaniline

Hazard: Toxic

Formula: C₆H₄ClFN₂O₂
Melting Pt: 146…147 °C
MDL Number: MFCD00052698
CAS Number: 104222-34-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 146-147°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR