N-Methyl-4-(trifluoromethoxy)aniline ≥97%

Supplier: Matrix Scientific

002468-1G 002468-5G
101776-114EA 50.77 USD
101776-114 101776-116
N-Methyl-4-(trifluoromethoxy)aniline ≥97%
N-Methyl-4-(trifluoromethoxy)aniline

Hazard: Toxic

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 96…98 °C (15 mmHg)
MDL Number: MFCD00052332
CAS Number: 41419-59-4

Order Now

Specification Test Results

Boiling point 96-98°/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR