2-Chloro-4-(trifluoromethyl)phenyl hydrazine ≥97%

Supplier: Matrix Scientific

002467-250MG 002467-1G
101776-110EA 46.15 USD
101776-110 101776-112
2-Chloro-4-(trifluoromethyl)phenyl hydrazine ≥97%
2-Chloro-4-(trifluoromethyl)phenyl hydrazine

Hazard: Toxic

Formula: C₇H₆ClF₃N₂
Boiling Pt: 80…85 °C (0.25 mmHg)
Melting Pt: 39…41 °C
MDL Number: MFCD00052916
CAS Number: 86398-98-3

Order Now

Specification Test Results

Boiling point 80-85°/0.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR