2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(trifluoromethyl)benzene-1-sulphonyl chloride

002466-5G 002466-1G
101776-108EA 80 USD
101776-108 101776-106
2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive

Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07 g/mol
Boiling Pt: 80…84 °C (1 mmHg)
Melting Pt: 35…39 °C
MDL Number: MFCD00052912
CAS Number: 175205-54-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 80-84°/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR