2-Chloro-4-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

002464-25G 002464-5G
101776-100EA 127.69 USD
101776-100 101776-098
2-Chloro-4-(trifluoromethyl)pyridine ≥98%
2-Chloro-4-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
Boiling Pt: 48…50 °C (15 mmHg)
Density: 1.411
MDL Number: MFCD00042224
CAS Number: 81565-18-6
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Boiling point 48-50°/15m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR