3-Chloro-4-(trifluoromethoxy)aniline ≥97%

Supplier: Matrix Scientific

002448-5G 002448-25G
101776-092EA 29.23 USD
101776-092 101776-094
3-Chloro-4-(trifluoromethoxy)aniline ≥97%
3-Chloro-4-(trifluoromethoxy)aniline

Hazard: Toxic/Irritant/Refrigerate

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Density: 1.454
MDL Number: MFCD00190126
CAS Number: 64628-73-5

Order Now

Specification Test Results

Density 1.454

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR