3-Fluoro-4-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

002444-1G 002444-5G
101776-088EA 40 USD
101776-088 101776-090
3-Fluoro-4-(trifluoromethyl)phenylacetonitrile
3-Fluoro-4-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅F₄N
MDL Number: MFCD00236305
CAS Number: 247113-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR