3-Fluoro-4-(trifluoromethyl)benzoyl chloride ≥97%
Supplier: Matrix Scientific
002437-5G
002437-1G
101776-082EA
89.23
USD
101776-082
101776-080
3-Fluoro-4-(trifluoromethyl)benzoyl chloride ≥97%
3-Fluoro-4-(trifluoromethyl)benzoyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₈H₃ClF₄O MW: 226.56 g/mol |
MDL Number:
MFCD00236284 CAS Number: 216144-68-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...