3-Fluoro-4-(trifluoromethyl)benzonitrile ≥97%
Supplier: Matrix Scientific
002435-5G
002435-25G
002435-100G
101776-074EA
30.77
USD
101776-074
101776-076
101776-078
3-Fluoro-4-(trifluoromethyl)benzonitrile ≥97%
3-Fluoro-4-(trifluoromethyl)benzonitrile
Hazard: Toxic/Irritant
Formula:
C₈H₃F₄N Melting Pt: 45…48 °C |
MDL Number:
MFCD00236280 CAS Number: 231953-38-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...