2-Fluoro-4-(trifluoromethyl)phenylacetonitrile ≥98%

Supplier: Matrix Scientific

002430-1G 002430-5G
101776-070EA 46.15 USD
101776-070 101776-072
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile ≥98%
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅F₄N
MDL Number: MFCD00061179
CAS Number: 239087-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR