2-Methyl-4,4,4-trifluorobutyric acid ≥95%

Supplier: Matrix Scientific

002411-1G 002411-5G
101776-052EA 58.46 USD
101776-052 101776-054
2-Methyl-4,4,4-trifluorobutyric acid ≥95%
2-Methyl-4,4,4-trifluorobutyric acid

Hazard: Corrosive

Formula: C₅H₇F₃O₂
MW: 156.11 g/mol
Boiling Pt: 172-175°C
MDL Number: MFCD00190640
CAS Number: 99783-23-0

Order Now

Specification Test Results

Boiling point 172-175°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR