1-Fluoro-3-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluoronitrobenzene

002389-25G 002389-100G
101776-038EA 21.54 USD
101776-038 101776-040
1-Fluoro-3-nitrobenzene ≥97%
1-Fluoro-3-nitrobenzene

Hazard: Toxic/Irritant

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 205 °C
Density: 1.325
MDL Number: MFCD00007196
CAS Number: 402-67-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 205°
Density 1.325
ND20 1.525

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR