4-Fluoro-3-nitroaniline ≥97%

Supplier: Matrix Scientific

002388-100G 002388-500G
101776-034EA 27.69 USD
101776-034 101776-036
4-Fluoro-3-nitroaniline ≥97%
4-Fluoro-3-nitroaniline

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD00007833
CAS Number: 364-76-1

Order Now

Specification Test Results

Melting point 96-98°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR