2-Fluoro-3-methylpyridine ≥97%

Supplier: Matrix Scientific

002386-25G 002386-100G
101776-030EA 29.23 USD
101776-030 101776-032
2-Fluoro-3-methylpyridine ≥97%
2-Fluoro-3-methylpyridine

Hazard: Flammable

Formula: C₆H₆FN
Boiling Pt: 148…150 °C
Density: 1.098
MDL Number: MFCD00041223
CAS Number: 2369-18-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 148-150°
Density 1.098


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR