2-Fluoro-3-methylbenzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-fluoro-3-methylbenzene

002382-1G 002382-5G
101776-026EA 126.15 USD
101776-026 101776-028
2-Fluoro-3-methylbenzyl bromide ≥98%
2-Fluoro-3-methylbenzyl bromide

Hazard: Corrosive

Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 168 °C (15 mmHg)
MDL Number: MFCD00042470
CAS Number: 151412-12-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 168°/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR