4-Fluoro-3-methylbenzyl bromide

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-1-fluoro-2-methylbenzene

002379-1G 002379-5G
101776-022EA 121.54 USD
101776-022 101776-024
4-Fluoro-3-methylbenzyl bromide
4-Fluoro-3-methylbenzyl bromide

Hazard: Corrosive

Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD01631425
CAS Number: 261951-70-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR