4-Fluoro-3-methylbenzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Fluoro-3-methylphenyl)methanamine

002378-5G 002378-25G
101776-018EA 53.85 USD
101776-018 101776-020
4-Fluoro-3-methylbenzylamine ≥97%
4-Fluoro-3-methylbenzylamine

Hazard: Corrosive

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD01631490
CAS Number: 261951-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR