4-Fluoro-3-methylbenzamide

Supplier: Matrix Scientific

002374-1G 002374-5G
101776-010EA 60 USD
101776-010 101776-012
4-Fluoro-3-methylbenzamide
4-Fluoro-3-methylbenzamide

Hazard: Irritant

Formula: C₈H₈FNO
MDL Number: MFCD01631489
CAS Number: 261945-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR