5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride ≥98%
Supplier: Matrix Scientific
002370-1G
002370-5G
101776-006EA
26.15
USD
101776-006
101776-008
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride ≥98%
5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₉H₆Cl₂O₂S₂ Melting Pt: 147…148 °C |
MDL Number:
MFCD00173797 CAS Number: 166964-33-6 |
Specification Test Results
Melting point | 147-148° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...