4-Fluoro-3-methylanisole ≥97%

Supplier: Matrix Scientific

002369-5G 002369-25G
101776-002EA 30.77 USD
101776-002 101776-004
4-Fluoro-3-methylanisole ≥97%
4-Fluoro-3-methylanisole

Hazard: Irritant

Formula: C₈H₉FO
Boiling Pt: 174…177 °C
Density: 1.07
MDL Number: MFCD00042293
CAS Number: 2338-54-7

Order Now

Specification Test Results

Boiling point 174-177°
Density 1.07
ND20 1.492


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR