4-Fluoro-m-toluidine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-methylaniline, 5-Amino-2-fluorotoluene

002368-25G 002368-100G
101775-998EA 26.15 USD
101775-998 101776-000
4-Fluoro-m-toluidine ≥97%
4-Fluoro-m-toluidine

Hazard: Toxic

Formula: C₇H₈FN
MW: 125.15 g/mol
Melting Pt: 35…37 °C
MDL Number: MFCD00025294
CAS Number: 452-69-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 35-37°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR